missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Forskolin

Product Code. 18782764
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18782764 10 mg 10mg
30001193 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18782764 Supplier R&D Systems™ Supplier No. 1099/10
  • ONLINE EXCLUSIVE PRICE
    263.96 €
    235.20 € / 10mg
    Save 28.76 €
    10% Off
View all promotions
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Adenylyl cyclase activator

Forskolin, or coleonol, is a cell-permeable, potent, reversible and rapid activator of adenylyl cyclase, an enzyme that converts ATP to cAMP and pyrophosphate (EC50 = 0.5 μM). In combination with 5 other compounds, forskolin enables chemical reprogramming of mouse embryonic fibroblasts to pluripotent stem cells, without genetic factors: Valproic acid, sodium salt (Cat. No. 2815), RepSox (Cat. No. 3742), Tranylcypromine hydrochloride (Cat. No. 3852), CHIR 99021 (Cat. No. 4423) and 3-Deazaneplanocin A hydrochloride (Cat. No. 4703). In combination with IBMX (Cat. No. 2845), forskolin induces neural differentiation of mesenchymal stem cells by downregulating neuron restrictive silencer factor. In a rat model of Huntington's disease-like neurodegeneration, forskolin also exhibits neuroprotective effects. Hypotensive and vasodilatory agent.

Water-soluble analog, NKH 477 (Cat. No. 1603), and Forskolin synthesized to Ancillary Material Grade also available.

For more information about how Forskolin may be used, see our protocol: Transdifferentiating Fibroblasts into Neurons.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66575-29-9
Molecular Formula C22H34O7

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 410.51
Purity >98%
Molecular Weight (g/mol) 410.507
Chemical Name or Material (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-5-(Acetyloxy)-3-ethenyldodecahydro-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1H-naphtho[2,1-b]pyran-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.