missing translation for 'onlineSavingsMsg'
Learn More

Forskolin, 98+%, Thermo Scientific™

Forskolin, CAS # 66575-29-9, is a diterpenoid compound isolated from Indian Coleus that acts as a strong activator of the adenylate cyclase system.

Brand:  Thermo Scientific Alfa Aesar J63292.MF

Quantity 50mg

 View more versions of this product

Product Code. 15445559

  • 823.00 € / 50mg
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description

• Forskolin is a diterpenoid and cyclic ketone, found naturally as a plant metabolite

• It is a protein kinase A agonist and a cyclic ketone • Forskolin is an antihypertensive and muscle relaxant; it can also promote the release of pituitary hormones and prevent platelet aggregation

Application

• Forskolin assists in neuron differentiation in vitro

• Useful for studies of cell metabolism and development; there is some evidence of its usefulness in the study of glaucoma

• It is intended for cell culture and biochemistry applications

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

66575-29-9
410.507
OHCQJHSOBUTRHG-KGGHGJDLSA-N
47936
[(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate
C22H34O7
MFCD00082317
forskolin, colforsin, coleonol, colforsina, colforsine, colforsinum, boforsin, colforsine french, colforsinum latin, colforsina spanish
CHEBI:42471
CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O

Specifications

66575-29-9
MFCD00082317
1692716
forskolin, colforsin, coleonol, colforsina, colforsine, colforsinum, boforsin, colforsine french, colforsinum latin, colforsina spanish
OHCQJHSOBUTRHG-KGGHGJDLSA-N
[(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate
47936
410.51
Forskolin
C22H34O7
50mg
14,2476
Soluble in ethanol,dimethylsulfoxide and chloroform; Insolule in water.
CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O
410.507
CHEBI:42471
≥98%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.