missing translation for 'onlineSavingsMsg'
Learn More

Forskolin, (from Coleus forskohlii), MP Biomedicals™

Product Code. 11446071 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446071 1 mg 1mg
11456071 5 mg 5mg
11466071 10 mg 10mg
11476071 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11446071 Supplier MP Biomedicals Supplier No. 0219066901
  • ONLINE EXCLUSIVE PRICE
    41.00 €
    38.85 € / 1mg
    Save 2.15 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 66575-29-9
Molecular Formula C22H34O7
Molecular Weight (g/mol) 410.507
MDL Number MFCD00082317
InChI Key OHCQJHSOBUTRHG-KGGHGJDLSA-N
Synonym forskolin, colforsin, coleonol, colforsina, colforsine, colforsinum, boforsin, colforsine french, colforsinum latin, colforsina spanish
PubChem CID 47936
ChEBI CHEBI:42471
IUPAC Name [(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate
SMILES CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O

Specifications

Color White
Assay Percent Range 0.9775
Quantity 1 mg
Formula Weight 410.5
Physical Form Powder
Chemical Name or Material Forskolin, from Coleus forskohlii
compliance-icons
Product Identifier
  • Forskoli
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.