missing translation for 'onlineSavingsMsg'
Learn More

Foscarnet Sodium Hexahydrate, TRC

Product Code. 17950735
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17950735 5 g 5g
17923245 100 mg 100mg
17942295 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17950735 Supplier Toronto Research Chemicals Supplier No. TRCF7274005G
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34156-56-4
Molecular Formula C O5 P . 3 Na . 6 H2 O
Molecular Weight (g/mol) 300.04
Synonym Foscarnet sodium hexahydrate, Trisodium phosphonatoformate hexahydrate, Phosphinecarboxylic acid, 1,1-dihydroxy-, 1-oxide, sodium salt, hydrate (1:3:6), Formic acid, phosphono-, trisodium salt, hexahydrate (8CI), Phosphinecarboxylic acid, dihydroxy-, oxide, trisodium salt, hexahydrate (9CI), Trisodium phosphonoformate hexahydrate
IUPAC Name trisodium;phosphonatoformate;hexahydrate
SMILES O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]C(=O)P(=O)([O-])[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/CH3O5P.3Na.6H2O/c2-1(3)7(4,5)6;;;;;;;;;/h(H,2,3)(H2,4,5,6);;;;6*1H2/q;3*+1;;;;;;/p-3
Formula Weight 299.981
Chemical Name or Material Foscarnet sodium hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.