missing translation for 'onlineSavingsMsg'
Learn More

Fosetyl Aluminum-d15 (Technical Grade), TRC

Product Code. 30394708
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394708 25 mg 25mg
30399330 10 mg 10mg
30403181 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30394708 Supplier Toronto Research Chemicals Supplier No. TRCF72745225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fosetyl Aluminum-d15
Molecular Formula C6 D15 H3 Al O9 P3
Quantity 25 mg
InChI Formula InChI=1S/3C2H7O3P.Al/c3*1-2-5-6(3)4;/h3*6H,2H2,1H3,(H,3,4);/q;;;+3/p-3/i3*1D3,2D2;
SMILES [2H]C([2H])([2H])C([2H])([2H])OP(=O)O[Al](OP(=O)OC([2H])([2H])C([2H])([2H])[2H])OP(=O)OC([2H])([2H])C([2H])([2H])[2H]
IUPAC Name tris(1,1,2,2,2-pentadeuterioethoxyphosphonoyloxy)alumane
Molecular Weight (g/mol) 369.197
Formula Weight 369.0921
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.