missing translation for 'onlineSavingsMsg'
Learn More

Fosetyl Aluminum (Technical Grade), TRC

Product Code. 30135532
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135532 1 g 1g
30135831 100 mg 100mg
17973965 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135532 Supplier Toronto Research Chemicals Supplier No. TRCF7274501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39148-24-8
Molecular Formula 3 C2 H6 O3 P . Al
Molecular Weight (g/mol) 354.1
Synonym Phosphonic acid, monoethyl ester, aluminum salt (9CI), Aliette, Aliette 80WG, Aliette Flash, Aliette WDG, Aliette WP, Alliete, Aluminum tris(O-ethylphosphonate), Aluminum tris(ethyl phosphite), Chipco Aliette Signature, Chipco Signature, EXP 1659, Efal, Efal 50, Efosite Al, Ephal, Fosethyl Al, Fosetyl Al, Fosetyl aluminum, LS 74-783, Phosethyl Al, Signature 80 WDG, Sinophos, Yilinlu, Fosetyl Aluminium Salt, Fosetyl Aluminium
IUPAC Name aluminum;ethoxyphosphinate
SMILES [Al+3].CCOP(=O)[O-].CCOP(=O)[O-].CCOP(=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/3C2H7O3P.Al/c3*1-2-5-6(3)4;/h3*6H,2H2,1H3,(H,3,4);/q;;;+3/p-3
Formula Weight 353.9979
Chemical Name or Material Fosetyl-aluminium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.