missing translation for 'onlineSavingsMsg'
Learn More

Fosinopril Sodium Salt, TRC

Product Code. 30135751
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135751 100 mg 100mg
30133142 25 mg 25mg
30011348 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135751 Supplier Toronto Research Chemicals Supplier No. TRCF727800100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88889-14-9
Molecular Formula C30 H45 N O7 P . Na
Molecular Weight (g/mol) 585.64
Synonym (4S)-4-Cyclohexyl-1-[[(R)-[(1S)-2-methyl-1-(1-oxopropoxy)propoxy](4-phenylbutyl)phosphinyl]acetyl]-L-proline, sodium salt, [1[S*(R*)],2alpha,4beta]-4-Cyclohexyl-1-[[[2-methyl-1-(1-oxopropoxy)propoxy](4-phenylbutyl)phosphinyl]acetyl]-L-proline, sodium salt, Acecor, Eosinopril, Fosinopril sodium, Monopril, SQ 28555, Secorvas, Staril
IUPAC Name sodium;(2S,4S)-4-cyclohexyl-1-[2-[[(1S)-2-methyl-1-propanoyloxypropoxy]-(4-phenylbutyl)phosphoryl]acetyl]pyrrolidine-2-carboxylate
SMILES [Na+].CCC(=O)O[C@@H](O[P@](=O)(CCCCc1ccccc1)CC(=O)N2C[C@@H](C[C@H]2C(=O)[O-])C3CCCCC3)C(C)C

Specifications

InChI Formula InChI=1S/C30H46NO7P.Na/c1-4-28(33)37-30(22(2)3)38-39(36,18-12-11-15-23-13-7-5-8-14-23)21-27(32)31-20-25(19-26(31)29(34)35)24-16-9-6-10-17-24;/h5,7-8,13-14,22,24-26,30H,4,6,9-12,15-21H2,1-3H3,(H,34,35);/q;+1/p-1/t25-,26+,30+,39-;/m1./s1
Quantity 100 mg
Formula Weight 585.2831
Shipping Condition Room Temperature
Chemical Name or Material Fosinopril sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.