missing translation for 'onlineSavingsMsg'
Learn More

Fostemsavir Tris, MedChemExpress

Product Code. 16449107
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16449107 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16449107 Supplier Medchem Express Supplier No. HY15440B
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Fostemsavir Tris (BMS-663068 (Tris)) is the phosphonooxymethyl prodrug of BMS-626529. Fostemsavir Tris (BMS-663068 (Tris)) is a novel attachment inhibitor that targets HIV-1 gp120 and prevents its binding to CD4+ T cells.

Fostemsavir Tris (BMS-663068 (Tris)) is the phosphonooxymethyl prodrug of BMS-626529. Fostemsavir Tris (BMS-663068 (Tris)) is a novel attachment inhibitor that targets HIV-1 gp120 and prevents its binding to CD4+ T cells.

Specifications

Chemical Name or Material Fostemsavir Tris
CAS 864953-39-9
Color White
Purity Grade Notes Research
Molecular Formula C29H37N8O11P
Quantity 5 mg
Synonym BMS-663068 Tris
Solubility Information DMSO : 125 mg/mL (177.40 mM; Need ultrasonic) ∣H2O : 100 mg/mL (141.92 mM; Need ultrasonic)
SMILES O=C(N1CCN(C(C2=CC=CC=C2)=O)CC1)C(C3=CN(COP(O)(O)=O)C4=C3C(OC)=CN=C4N5N=C(C)N=C5)=O.OCC(CO)(N)CO
Molecular Weight (g/mol) 704.62
Formula Weight 704.62
Percent Purity 99.94%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.