missing translation for 'onlineSavingsMsg'
Learn More

Fostriecin sodium salt, Tocris Bioscience™

Product Code. 18055352
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18055352 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18055352 Supplier Tocris Bioscience Supplier No. 1840/50U

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent PP2A and PP4 inhibitor

Potent inhibitor of protein phosphatase types 2A (PP2A) and 4 (PP4) with IC50 values of 1.5 nM and 3 nM respectively. Also exhibits weaker inhibition of topoisomerase II (IC50 = 40 μM) and protein phosphatase type 1 (PP1) (IC50 = 131 μM) with no apparent inhibition of protein phosphatase type 2B (PP2B). Active in vivo. Originally identified as an antitumor antibiotic from Streptomyces pulveraceous.
TRUSTED_SUSTAINABILITY

Specifications

CAS 87860-39-7
Molecular Formula C19H26NaO9P
Quantity 50 μg
Synonym fostriecin sodium salt
InChI Key XBUIKNRVGYFSHL-SOAGNPLSSA-M
SMILES CC(C=CC1CC=CC(=O)O1)(C(CC(C=CC=CC=CCO)O)OP(=O)(O)[O-])O.[Na+]
IUPAC Name sodium;[(1Z,3R,4R,6R,7E,9E,11E)-3,6,13-trihydroxy-3-methyl-1-[(2R)-6-oxo-2,3-dihydropyran-2-yl]trideca-1,7,9,11-tetraen-4-yl] hydrogen phosphate
Molecular Weight (g/mol) 452.372
PubChem CID 124080892
Formula Weight 452.37
Percent Purity >98%
Chemical Name or Material Fostriecin sodium salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.