missing translation for 'onlineSavingsMsg'
Learn More

FPTQ, MedChemExpress

Product Code. 16495717
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495717 5 mg 5mg
16479546 50 mg 50mg
16472978 10 mg 10mg
16462566 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16495717 Supplier Medchem Express Supplier No. HY100382
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress FPTQ is potent mGluR1 antagonist with IC50 values of 6 nM and 1.4 nM for human and mouse mGluR1 respectively. FPTQ has anti-oxidant and anti-inflammatory effects in vitro and in vivo.

FPTQ is potent mGluR1 antagonist with IC50 values of 6 nM and 1.4 nM for human and mouse mGluR1 respectively. FPTQ has anti-oxidant and anti-inflammatory effects in vitro and in vivo.

Specifications

Chemical Name or Material FPTQ
CAS 864863-72-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H12FN5
Quantity 5 mg
Solubility Information DMSO : 33.33 mg/mL (109.17 mM; Need ultrasonic)
SMILES FC(N=CC=C1)=C1N2N=NC(C3=CC4=C(C=C3)N=CC=C4)=C2C
Molecular Weight (g/mol) 305.31
Formula Weight 305.31
Percent Purity 99.88%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.