missing translation for 'onlineSavingsMsg'
Learn More

Skytec™ FRBH Boilersuit

Product Code. 31702843
Click to view available options
Size:
X-Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Unit Size:
Pack of 10

Product Code. 31702843

Brand: Skytec™ SKC030100EN

Please to purchase this item. Need a web account? Register with us today!

Protective boilersuit designed to provide chemical resistance along with protection from heat, flame, and electrostatic hazards, while ensuring comfort for the wearer.

  • Offers protection against heat, flame and electrostatic hazards
  • Double storm facing velcro closure for increased protection
  • Ring-pull zip for easy donning and doffing when wearing gloves

Specifications

Product Type Chemical Protective Clothing
Size 3X-Large
Gender Unisex
Material PVC
Color Green
Disposable Reusable
Waist Style Loose
Ankle Style Elastic
Cuff Style Open
Neck Style Hooded
Hood Type Attached
Closure Type Zipper
Seam Type Stitched, Taped
Sterility Non-sterile
Resistance Anti-static, Particle, Liquid, Chemicals, Flame
Certifications/Compliance EN 1149-5:2018 or - EN 11612:2015 A1,B1,C1 or - EN 14605:2005 + A1:2009 Type 3,Type 4
For Use With (Application) Public Works, Transportation of Fuels, Waste Collection and Sorting, Chemical Handling and Containment
Weight (Metric) 1440 g
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.