Learn More
Hach Lange™ Free Chlorine Reagent Set
Description
Hach Lange Free Chlorine Reagent Set, Range: 0.02 to 5.00 mg/L Cl2, For Use With: Free Chlorine Test, Includes: DPD Indicator, Free Chlorine Indicator and Buffer Solutions, Parameter Specifications: Chlorine, Test Method: 10059, DPD
Specifications
Specifications
| For Use With (Application) | Free Chlorine Test |
| Includes | DPD Indicator, Free Chlorine Indicator and Buffer Solutions |
| Parameter Specifications | Chlorine |
| Range | 0.02 to 5.00 mg/L Cl2 |
| Test Method | 10059, DPD |
| Type | Free Chlorine Reagent Set |