missing translation for 'onlineSavingsMsg'
Learn More

Euro Protection™ Freedite High-Cut Safety Shoes

Product Code. 15795615
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15795615 39 1 pair
15765615 36 1 pair
15775615 37 1 pair
15785615 38 1 pair
15705625 40 1 pair
15715625 41 1 pair
15725625 42 1 pair
15735625 43 1 pair
15745625 44 1 pair
15755625 45 1 pair
15765625 46 1 pair
15775625 47 1 pair
15785625 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
This item is not returnable. View return policy
Product Code. 15795615 Supplier Euro Protection™ Supplier No. 9FREH39
  • ONLINE EXCLUSIVE PRICE
    66.15 €
    32.97 € / 1 pairs
    Save 33.18 €
    50% Off
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Euro Protection™ Freedite High-Cut Safety Shoes are padded for a comfortable, snug fit and feature a footbridge for rigidity and a composite toe cap for safety.

  • Footbridge for torsional rigidity.
  • Padding for snug fit.
  • Also available with an electrical hazards resistant outsole on request (Reference: 9FEHH).
  • Full grain leather upper, reflective trim at the back.
  • Breathable 3D mesh lining.
  • Removable, textile and foam insole.
  • Composite toe cap.
  • Non-metallic (composite fabric) puncture resistant sole.
  • Injected polyurethane midsole.
  • Polyurethane outsole.

Specifications

Color Black
Liner Material Breathable 3D mesh lining
Material Leather
Size 39
Sole Features Removable, textile and foam insole, non-metallic (composite fabric) puncture-resistant sole, injected polyurethane midsole, polyurethane outsole.
Sole Material Polyurethane, Composite Fabric
Style High
Toe Material Composite
Weight (Metric) 602 g per shoe in size 8
Certifications/Compliance (Europe) EN ISO 20345 : 2011
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.