missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Freedom™ ExpiCHO-S™ Kit

Product Code. 30283903 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30283903 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30283903 Supplier Gibco™ Supplier No. A46847
Make Your Process Development Count
Use promo code 27978
View all promotions
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An easy-to-use, beginning-to-end product for cloning and expression of recombinant proteins in Chinese Hamster Ovary (CHO)-derived suspension culture. The Gibco™ Freedom™ ExpiCHO-S™ Kit includes components for transfection, expression, clone creation, and stable cell line selection, all in a conveniently packaged kit.

  • Seamless transition from transient to stable clones – all in one cell line
  • Regulatory-friendly cGMP-banked cells
  • Dual vector system provides ability to control heavy and light chain ratios
  • Ability to achieve titers of 3 to 5 g/L in fed batch cultures
  • Short development timeline with less than 5 months needed to get from gene of interest to clone
  • Access to technical support and consultation
  • Flexible, royalty-free commercial licensing options

Order Info

Kit components ship separately

TRUSTED_SUSTAINABILITY

Specifications

Cell Line ExpiCHO-S cells, cGMP Banked
For Use With (Application) Protein Production, Stable Cell Line Development
Quantity 1 Kit
Content And Storage • Gibco ExpiCHO Expression Medium, 2–8°C, protect from light
• ExpiCHO-S cells (cGMP-banked), liquid nitrogen
• One Shot TOP10 Chemically Competent E. coli, –80°C
• Freedom pCHO 3.0 Expression vectors, –5 to –20°C

Delivery Type Transfection
Expression System Mammalian

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.