missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ FreeStyle™ MAX Reagent

Product Code. 10489684
Click to view available options
Quantity:
1 mL
15 mL
10 x 15 mL
Unit Size:
15mL
1mL
Pack of 10
This item is not returnable. View return policy

Product Code. 10489684

Brand: Gibco™ 16447750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Animal origin-free formulation for transfecting plasmid DNA into eukaryotic cells that can easily be scaled up to produce large amounts of recombinant proteins

  • Allows highest expression levels and transfection rates with lowest cytotoxicity in bio-production applications
  • Formulated for use with:
  • FreeStyle 293-F Cells (suspension human embryonal kidney cells, Cat. No. R790-07) in serum-free FreeStyle 293 Expression Medium (Cat. No. 12338-018)
  • FreeStyle CHO-S Cells (suspension Chinese Hamster Ovary cells, Cat. No. R800-07) in serum-free FreeStyle CHO Expression Medium (Cat. No. 12651-014)
  • DG44 Cells (DHFR– suspension CHO cells) in CD DG44 Medium with 8 mM L-glutamine and 18 mL/L 10% Pluronic F-68 (Cat. No. A11000-01)
  • FreeStyle™ MAX Reagent is a proprietary, animal origin–free formulation for transfecting plasmid DNA into eukaryotic cells that can easily be scaled up to produce large amounts of recombinant proteins
  • Intended for use with the FreeStyle™ MAX 293 Expression System (Cat. No. K9000-10) and the FreeStyle™ MAX CHO Expression System (Cat. No. K9000-20).
TRUSTED_SUSTAINABILITY

Specifications

Classification Animal Origin-Free
Content And Storage Each kit contains 10 units. Each unit contains one vial of reagent. Store at +4°C. FreeStyle™ MAX is guaranteed stable for 6 months when stored properly.
Cell Type Established Cell Lines
Format Tube
Sample Type Plasmid DNA
Transfection Technique Lipid-based Transfection
For Use With (Application) Protein Expression
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line Freestyle
Product Type Transfection Reagent
Quantity 10 x 15 mL
Serum Compatible Yes
Shipping Condition Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.