Learn More
Gardner Denver Welch™ Kit 6 Freeze dryer adapter kit, for Chemvac & Vacuum concentrator Adapter kit, DN 25 KF
Product Code. 13226349 Shop All Welch Ilmvac Products
Change view
Click to view available options
For Use With (Equipment):
Rotary vane pump DN 25 KF
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
13226349 Rotary vane pump DN 25 KF Each
1 options
Product Code. 13226349 Supplier Gardner Denver Welch™ Supplier No. 330045
  • ONLINE EXCLUSIVE PRICE
    174.00 €
    170.10 € / Pack of 1
    Save 3.90 €
    2% Off
 Restricted Item
Estimated Shipment: 04-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For rotary vane pump DN 25 KF
Vacuum concentrator kit includes:
  • KF25 flange adapter
  • 1.5m PVC vacuum hose
  • Clamps and centring rings
TRUSTED_SUSTAINABILITY
Specifications
Type Freeze Dryer and Vacuum Concentrator Adapter Kit
For Use With (Equipment) Rotary vane pump DN 25 KF