Learn More
Eppendorf™ Freezer Rack: CryoCube™ F740 (3-comp., MAX), 2 in/53 mm
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Brand: Eppendorf™ 6001071210
194.66 EUR valid until 2025-08-22
Use promo code "24148" to get your promotional price.
Description
Freezer Rack: CryoCube™ F740 series (3-compartment, MAX), 2 in/53 mm, side access, stainless steel- Rack for 53 mm/ 2 in boxes- Max. box size [mm]: 136 x 136- Side access rack (MAX)- Ext. dimensions of rack (DxWxH) [mm]: 569 x 139 x 444 (22.4 x 5.5 x 17.5 in)- Max. box number per rack: 32 (4 x 8)- Material: Stainless steel- For ULT freezer: CryoCube™ F740 series 3-compartment (Max. 18x racks)- For ULT freezer: CryoCube F740 series 3-compartment (Max. 576x boxes)

Specifications
Freezer Rack | |
56.9 x 13.9 x 44.4 cm/5.5 x 22.4 x 17.5 in. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.