Learn More
Eppendorf™ Freezer Rack: CryoCube™ FC660 series, 2 in/53 mm, side access, stainless steel
Description
Freezer Rack: CryoCube™ FC660 series, 2 in/53 mm, side access, stainless steel- Rack for 53 mm/2 in boxes- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 141 x 141 x 721 (5.6 x 5.6 x 28.4 in)- Max. box number per rack: 13 (13 x 1)- Stainless steel- For ULT freezer: CryoCube™ FC660/ FC660h (max. 40 racks)- For ULT freezer: CryoCube™ FC660/ FC660h (max. 520 boxes)
Specifications
Specifications
| Product Type | Freezer Rack |
| Material | Stainless Steel |