Learn More
Eppendorf™ Freezer Rack: Innova™ U101, CryoCube™ F101h, 2 in/53 mm
Product Code. 15773265
Change view
Click to view available options
Product Type:
Freezer Rack
Unit Size:
Each
Product Code. Product Type Unit Size
15773265 Freezer Rack Each
1 options
Product Code. 15773265 Supplier Eppendorf™ Supplier No. 6001062210
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Freezer Rack: Innova™ U101, CryoCube™ F101h, 2 in/53 mm, drawer, stainless steel- Rack for 53 mm/2 in boxes- Max. box size [mm]: 136 x 136- Drawer rack- Ext. dimensions of rack (DxWxH) [mm]: 308 x 145 x 290 (12.1 x 5.7 x 11.4 in)- Max. box number per rack: 10 (5 x 2)- Stainless steel- For ULT freezer: CryoCube F101h/ Innova U101 (max. 6 racks)- For ULT freezer: CryoCube F101h/ Innova U101 (max. 60 boxes)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Freezer Rack