missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ibidi Freezing Medium Classic
Click to view available options
Reconstituted Volume:
20 mL
120 mL
Unit Size:
Each
Description
- A cell freezing medium for standard cell lines
- with Bovine Serum Albumin
- 5 x 20 mL or 120 mL
Specifications
Specifications
| Product Type | Freezing Medium |
| With Additives | Bovine Serum Albumin (BSA) |
| Quantity | 5 |
| Reconstituted Volume | 20 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction