missing translation for 'onlineSavingsMsg'
Learn More

Labconco FreeZone Stoppering Tray Dryer

Product Code. 17980190 Shop All Labconco Corporation Products
Change view
Click to view available options
Includes:
6-Port Manifold
Isolation Valve
Stoppering Tray Dryer
Unit Size:
1 unit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
17980190 Isolation Valve 1 unit
17960180 Stoppering Tray Dryer 1 unit
17980180 6-Port Manifold 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17980190 Supplier Labconco Supplier No. 794801290
  • ONLINE EXCLUSIVE PRICE
    38620.00 €
    35489.49 € / Pack of 1
    Save 3130.51 €
    8% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

FreeZone Stoppering Tray Dryers lyophilize moderate to large loads of vials or serum bottles and stopper them under vacuum. A pneumatic mechanism stoppers containers under vacuum on three large adjustable processing shelves.

  • A pneumatic mechanism stoppers containers under vacuum on three large adjustable processing shelves, 16.5 x 12 in. each.
  • An HCFC/CFC-free refrigeration system and electric heater cool and heat fluid medium that is circulated through the shelves to maintain shelf temperatures within ±1° C.
  • Lyo-Works™ OS lets the user set up to 30 programs, with up to 16 steps each; record, chart and store data; and receive email alerts.
  • The tray dryer connects to a FreeZone Console Freeze Dryer that provides a real time display of the tray dryer’s parameters on a large capacitive touchscreen.
  • For use with any FreeZone Freeze Dryer manufactured after July 2017.

Specifications

Type Stoppering Tray Dryer
Capacity (Metric) 6 L, 12 L, or 18 L Freeze Dryer
No. of Shelves 3
Temperature Range (Metric) -40°C to +40°C
Material Stainless Steel
Includes Isolation Valve
Plug Type All except North America
Voltage 230 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.