missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies FreSR™-S

Product Code. 15567540
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15567540 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15567540 Supplier STEMCELL Technologies Supplier No. 05859
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Animal component-free medium for freezing ES and iPS cells as single cells

  • FreSR™-S is a defined, serum-free, and animal component-free medium for the cryopreservation of human pluripotent stem cells (hPSCs) as single cells
  • This complete and ready-to-use medium is recommended for hPSCs cultured in mTeSR™1, mTeSR™ Plus, TeSR™2, or TeSR™-E8™
  • Cryopreserved hPSCs should be stored at -135°C (liquid nitrogen) or colder

Specifications

Cell Type Pluripotent Stem Cells
For Use With (Application) Cryopreservation
Quantity 50 mL
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.