missing translation for 'onlineSavingsMsg'
Learn More

Vileda Professional™ Roll-O-Matic™ Sponge Mop Refills

Product Code. 17882456
Change view
Click to view available options
Material:
CE Universal Stainless Steel
Microfiber, Stainless Steel
Polyester with Stainless Steel Channel
Polyester, Stainless Steel
Stainless Steel
Width (English):
10 in.
14 in.
Description:
“Clean”
“Disinfect”
“Universal”
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 12
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Width (English) Description Sterility unitSize
17882456 Microfiber, Stainless Steel 14 in. “Clean” Non-sterile Pack of 12
17822506 Polyester with Stainless Steel Channel 14 in. “Disinfect” Non-sterile Pack of 12
17802466 Microfiber, Stainless Steel 10 in. “Clean” Non-sterile Pack of 12
17842456 Polyester, Stainless Steel 10 in. “Disinfect” Non-sterile Pack of 12
17822456 Stainless Steel 10 in. “Universal” Sterile Pack of 12
17832476 CE Universal Stainless Steel 14 in. “Universal” Sterile Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17882456 Supplier Vileda Professional™ Supplier No. 119657
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Vileda Professional Roll-O-Matic™ sponge mop refills are made in the USA with Advanced Technology foam. They are manufactured specifically for the controlled environment industry, and allow 25% more liquid absorption and 2X more liquid release than competing foams.

Vileda Professional Roll-O-Matic™ sponge mop refills are made in the USA with Advanced Technology foam. They are manufactured specifically for the controlled environment industry, and allow 25% more liquid absorption and 2X more liquid release than competing foams.

All Vileda Professional brand refills are ideal for use with the "Pull and Lift" technique most commonly used in controlled environments. They are ideal for general use, absorbing spills, applying disinfectant, and cleaning particulates.

  • Fits all original and self-wringing Roll-O-Matic™ mops
  • 10- or 14-inch options for both stainless steel and galvanized steel handles
  • Completely autoclavable and resistant to gamma and ETO sterilization
  • Individually packaged and lot number controlled
  • Certificate of conformance and technical data available upon request
  • Sterile versions validated to 10-6 per ISO 11137, individually double bagged with lot number and indicator dot, and supplied with certificate of sterility upon request

Specifications

Product Type Roller Mop Refill
Color Blue, White
Sterility Non-sterile
Autoclavable Autoclavable
Width (English) 14 in.
Quantity 12/Cs.
For Use With (Equipment) Original and Self Wringing Roll-O-matic Roller Mops
Description “Clean”
For Use With (Application) For Tear Free Cleaning of Deep Contaminants and Disinfectant Residues
Material Microfiber, Stainless Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.