missing translation for 'onlineSavingsMsg'
Learn More

Vileda Professional™ CE Dip-N-Go Sieve

Product Code. 17862486
Change view
Click to view available options
Color:
Gray
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17862486 Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17862486 Supplier Vileda Professional™ Supplier No. 141928
  • ONLINE EXCLUSIVE PRICE
    213.00 €
    183.21 € / Pack of 1
    Save 29.79 €
    13% Off
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dip-N-Go Sieve for flat mops. Can be used on either CE Bucket Trolley or CE Pre-Prepared Trolley

  • For use when high disinfectant times are needed or deep wringing is not required
  • Can be used with either CE Pre-Prepared trolley or CE Bucket Systems Trolley
  • Stainless Steel
  • Autoclavable

Specifications

Product Type CE Dip-N-Go Sieve
Material Stainless Steel
Autoclavable Autoclavable
Color Gray
Dimensions 21 x 7 x 2 in. (54 x 17 x 5 cm)
For Use With (Equipment) CE Bucket Trolley
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.