missing translation for 'onlineSavingsMsg'
Learn More

Vileda Professional™ CE Bucket Trolley

Product Code. 17842486
Change view
Click to view available options
Color:
Gray
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17842486 Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17842486 Supplier Vileda Professional™ Supplier No. 141926
  • ONLINE EXCLUSIVE PRICE
    3325.00 €
    2875.98 € / Pack of 1
    Save 449.02 €
    13% Off
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for use in controlled environments where maximum contamination control needed

The trolleys are electro-polished stainless steel with air tight connections and smooth weldings. This ensures excellent biological control while still being easy to clean and full autoclavable. Trolleys offer ample space for all utensils needed for effective cleaning and are fully customizable to meet your exact needs with optional accessories.

  • Two stainless steel mop handle holders
  • Support bar for Duo Press Wringer (sold separately) or Dip-N-Go Sieve (sold separately), Support Plate (sold separately) and commonly used industry wringers
  • Frame holders allow for storage of various sized mops
  • Rear area working station
  • Third bucket extension available turning double bucket system into triple bucket system (sold separately)
  • Autoclavable, extremely quiet cleanroom casters
  • Electro-polished for a contamination free surface
  • Color coded system within boxes, buckets and handles (all accessories sold separately)

Certifications

ISO Class 5.

Specifications

Product Type Double Bucket Trolley
Material Stainless Steel
Color Gray
Dimensions 77 x 71 x 95 cm (30.3 x 28 x 37.4 in.)
For Use With (Application) ISO 5, LAR, Production, Laboratory
For Use With (Equipment) Vileda Professional-FHP CE Dip-N-Go Sieve, Support Plate for CE Dip-N-Go Sieve, 25L Longish Bucket With Color Coding Clips, and Bucket Extension
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.