missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Frontier™ 5000 Multi-Pro Series Rotors and Adapters Rack

Product Code. 30130258 Shop All Ohaus Products
Click to view available options
Capacity:
24 x 1.5/2 mL
8 x 1.5 mL/2.0 mL
Unit Size:
Pack of 2
This item is not returnable. View return policy

Product Code. 30130258

Brand: OHAUS™ 83041030

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Offering flexibility to maximize sample preparation.

Frontier™ Multi-Pro Series centrifuge accessory.

  • Rack
  • 2 pack
TRUSTED_SUSTAINABILITY

Specifications

Type Rack
Material Polypropylene
For Use With (Equipment) 83041011
Disposable Reusable
Capacity 8 x 1.5 mL/2.0 mL
Autoclavable Not Autoclavable
Tube Size 1.5/2.0 mL
Diameter (Metric) Inside 11 mm
No. per Pack 2 Pieces
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.