Learn More
Abcam Fructose Assay Kit (Fluorometric)
Product Code. 32111249 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
Product Code. Quantity Unit Size
32111249 100 Tests 100 tests
1 options
Product Code. 32111249 Supplier Abcam Supplier No. ab241022100Test
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Assay Fructose Assay in 1h 15m in Biofluids, Cell/Tissue Extracts with Fructose Assay Kit AB241022. Sensitivity = 0.005 nmol/well. Cited in 1 publication. Use with Microplate reader.
Assay Fructose Assay in 1h 15m in Biofluids, Cell/Tissue Extracts with Fructose Assay Kit AB241022. Sensitivity = 0.005 nmol/well. Cited in 1 publication. Use with Microplate reader.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 100 Tests