missing translation for 'onlineSavingsMsg'
Learn More

Fructose-histidine, TRC

Product Code. 30179449
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179449 100 mg 100mg
30178932 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179449 Supplier Toronto Research Chemicals Supplier No. TRCF792590100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25020-13-7
Molecular Formula C12H19N3O7
Molecular Weight (g/mol) 317.3
Synonym (S)-1-[[1-Carboxy-2-(1H-imidazol-4-yl)ethyl]amino]-1-deoxy-D-fructose;D-1-[(1-Carboxy-2-imidazol-4-ylethyl)amino]-1-deoxy-fructose
SMILES O[C@@H]1COC(O)(CN[C@@H](Cc2cnc[nH]2)C(=O)O)[C@@H](O)[C@@H]1O

Specifications

Quantity 100 mg
Formula Weight 317.1223
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material Fructose-histidine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.