Learn More
American Educational Products Frufly — A Genetics Experience
Simulate genetics research with simulated fruit flies.
Brand:  American Educational Products R-100872
Product Code. 13193120
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Includes:
  • Teacher's manual
  • 15 plastic sheets with snap-apart features showing wild type, wingless, eyeless and bar eyes
Description
Description
Many principles of genetics revolve around the organism known as Drosophila melanogaster, commonly called the fruit fly, but handling these tiny organisms sometimes presents difficulties. We have solved that problem with our Frufly kit, which demonstrates genetic crosses with simulated fruit flies.
Observe and simulate the following:
  • Sex determination
  • Single-factor inheritance
  • Double-factor inheritance
  • Sex linkage
  • Dominance and recessiveness
  • Punnett square calculations