missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ FSEN 1

Product Code. 30040180
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040180 10 mg 10mg
30040214 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30040180 Supplier Tocris Bioscience™ Supplier No. 7941/10
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Induces ferroptosis; inhibits FSP1

FSEN 1 is a ferroptosis suppressor protein 1 (FSP1) inhibitor (IC50 = 313 nM). FSEN 1 sensitizes lung cancer cells and organoids to ferroptosis (EC50 = 69 nM) through inhibition of FSP1. Inhibits FSP1 CoQ1 oxidoreductase activity (IC50 = 688 nM).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(4-Bromophenyl)-6-((4-(4-methoxyphenyl)piperazin-1-yl)methyl)thiazolo[2,3-c][1,2,4]triazole
CAS 862808-01-3
Quantity 10 mg
Target Ferroptosis Inhibitors
Molecular Formula C22H22BrN5OS
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.