missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ FSEN 1
Description
FSEN 1 is a ferroptosis suppressor protein 1 (FSP1) inhibitor (IC50 = 313 nM). FSEN 1 sensitizes lung cancer cells and organoids to ferroptosis (EC50 = 69 nM) through inhibition of FSP1. Inhibits FSP1 CoQ1 oxidoreductase activity (IC50 = 688 nM).
Specifications
Specifications
| Chemical Name or Material | 3-(4-Bromophenyl)-6-((4-(4-methoxyphenyl)piperazin-1-yl)methyl)thiazolo[2,3-c][1,2,4]triazole |
| CAS | 862808-01-3 |
| Quantity | 50 mg |
| Target | Ferroptosis Inhibitors |
| Molecular Formula | C22H22BrN5OS |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?