missing translation for 'onlineSavingsMsg'
Learn More

FTI-277 hydrochloride, MedChemExpress

Product Code. 16486966
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16486966

Brand: Medchem Express HY15872A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress FTI-277 hydrochloride is an inhibitor of farnesyl transferase (FTase); a highly potent Ras CAAX peptidomimetic which antagonizes both H- and K-Ras oncogenic signaling. FTI-277 hydrochloride can inhibit hepatitis delta virus (HDV) infection.

FTI-277 hydrochloride is an inhibitor of farnesyl transferase (FTase)

Specifications

Chemical Name or Material FTI-277 hydrochloride
CAS 180977-34-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H30ClN3O3S2
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (206.58 mM; Need ultrasonic) ∣H2O : 50 mg/mL (103.29 mM; Need ultrasonic)
SMILES CSCC[C@@H](C(OC)=O)NC(C1=CC=C(NC[C@@H](N)CS)C=C1C2=CC=CC=C2)=O.[H]Cl
Molecular Weight (g/mol) 484.07
Formula Weight 484.07
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.