missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ FTY 720

Product Code. 18792168
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18792168 10 mg 10mg
18761508 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18792168 Supplier R&D Systems™ Supplier No. 6176/10
  • ONLINE EXCLUSIVE PRICE
    166.77 €
    149.10 € / 10mg
    Save 17.67 €
    10% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent S1P receptor agonist; also immunosuppressant

FTY 720 is an S1P receptor agonist; undergoes phosphorylation by sphingosine kinase 2, which then binds S1P1,3,4 and 5 receptors (EC50 values are 0.3 - 0.6 nM for S1P1, 4 and 5 and ∼3 nM for S1P3). Immunosuppressant; sequesters lymphocytes in lymph nodes, reducing autoimmune response. Suppresses the development of EAE (an in vivo model of MS). Orally bioavailable and blood-brain barrier permeable.
TRUSTED_SUSTAINABILITY

Specifications

CAS 162359-56-0
Quantity 10 mg
Product Line Tocris
Target Sphingosine-1-phosphate Receptor
Molecular Formula C19H33NO2.HCl
Formula Weight 343.93g/mol
Product Type Agonist

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.