missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Fuchsine base monohydrochloride

Product Code. 16491118
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491118 5 g 5g
16462349 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16491118 Supplier MedChemExpress Supplier No. HYB1539A
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fuchsine base (monohydrochloride) is a magenta dye, which is certified for use for acid-fast staining with carbol-fuchsin.

Specifications

Content And Storage 4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture)
Product Type Dye Reagents
Quantity 5 g
Shipping Condition Room temperature
CAS 632-99-5
Molecular Weight (g/mol) 337.85 g/mol
Synonym Magenta base monohydrochloride ∣ Basic Fuchsin monohydrochloride ∣ Rosaniline Base monohydrochloride
Target HBV
Solubility DMSO : 50 mg/mL (147.99 mM; Need ultrasonic) ∣ H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.