missing translation for 'onlineSavingsMsg'
Learn More

Fucoidan, Thermo Scientific™

Product Code. 17585535 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17585535 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17585535 Supplier Thermo Scientific Acros Supplier No. 467460010
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

Alpha Vector FUCOIDAN
CAS 9072-19-9
Color Light yellow to brown
Loss on Drying ≤10 %
Packaging Glass Bottle
MDL Number 00131109
Quantity 1 g
Physical Form Powder
Chemical Name or Material Fucoidan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.