missing translation for 'onlineSavingsMsg'
Learn More

(-)-Fucose, MedChemExpress

Product Code. 16439166
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16439166 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16439166 Supplier Medchem Express Supplier No. HYN1480

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress (-)-Fucose is classified as a member of the hexoses, plays a role in A and B blood group antigen substructure determination, selectin-mediated leukocyte-endothelial adhesion, and host-microbe interactions.

(-)-Fucose is classified as a member of the hexoses, plays a role in A and B blood group antigen substructure determination, selectin-mediated leukocyte-endothelial adhesion, and host-microbe interactions.

Specifications

Chemical Name or Material (-)-Fucose
CAS 2438-80-4
Color White
Purity Grade Notes Research
Molecular Formula C6H12O5
Quantity 100 mg
Synonym 6-Desoxygalactose L-(-)-Fucose L-Galactomethylose
Solubility Information H2O : 100 mg/mL (609.16 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (609.16 mM; Need ultrasonic)
SMILES O=C[C@H]([C@@H]([C@@H]([C@H](C)O)O)O)O
Molecular Weight (g/mol) 164.16
Formula Weight 164.16
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.