missing translation for 'onlineSavingsMsg'
Learn More

PURUS™ Full Finger Cut Resistant Glove Liners

Product Code. 30560017 Shop All Purus Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size
30560017 X-Large
30560014 Small
30560015 Medium
30560016 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30560017 Supplier PURUS™ Supplier No. GLFFXLCREU
  • ONLINE EXCLUSIVE PRICE
    2300.00 €
    1563.31 € / 200 pairs
    Save 736.69 €
    32% Off
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Glove liners designed to be worn under latex, PVC, or nitrile gloves to wick away moisture and prevent possible hand injuries.

  • Constructed of 13-gauge white nylon and HPPE
  • Effective underglove for all cleanroom requirements
  • Color-coded cuffs for easy size identification
  • Launderable for reuse

Specifications

Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Latex Free Yes
Material Nylon, HPPE
Product Type Full Glove Liners
Size X-Large
Sterility Non-sterile
Style Full-hand Mitt
For Use With (Application) Latex, PVC or Nitrile Gloves
Gender Unisex
Glove Cuff Color Red
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.