missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fulvestrant

Product Code. 16617042 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
Product Code. Quantity unitSize
16617042 1 g 1g
16627042 50 mg 50mg
16637042 250 mg 250mg
3 options
This item is not returnable. View return policy

Product Code. 16617042

Brand: Thermo Scientific Chemicals 465780010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129453-61-8
Molecular Formula C32 H47 F5 O3 S

Specifications

Quantity 1 g
Solubility (0.1 g in 10 mL Ethanol) Clear solution
Formula Weight 606.77
Physical Form Powder
Product Type Reagent
Purity ≥98.5% (HPLC)
InChI Key VWUXBMIQPBEWFH-WCCTWKNTSA-N
SMILES C[C@]12CC[C@H]3[C@H]([C@@H]1CC[C@@H]2O)[C@H](CCCCCCCCCS(=O)CCCC(F)(F)C(F)(F)F)CC1=CC(O)=CC=C31
IUPAC Name (1S,3aS,3bR,4R,9bS,11aS)-11a-methyl-4-[9-(4,4,5,5,5-pentafluoropentanesulfinyl)nonyl]-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1,7-diol
Molecular Weight (g/mol) 606.78
Chemical Name or Material Fulvestrant
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.