missing translation for 'onlineSavingsMsg'
Learn More

Fumaric acid, 99%

Product Code. 11424423
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
This item is not returnable. View return policy

Product Code. 11424423

Brand: Thermo Scientific Alfa Aesar A10976.0E

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-17-8 | C4H4O4 | 116.07 g/mol

Fumaric acid is used as a food additive and acid regulator. It is used in the production of polyester resins and polyhydric alcohols. It is also used as a mordant for dyes, paint additives, coating additives, plasticizers, processing aids and specific to petroleum production. Further, it is used in dentifrices as a stain remover and to make other chemicals. In addition to this, it is used in rosin esters, drying oils, printing inks and flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fumaric acid is used as a food additive and acid regulator. It is used in the production of polyester resins and polyhydric alcohols. It is also used as a mordant for dyes, paint additives, coating additives, plasticizers, processing aids and specific to petroleum production. Further, it is used in dentifrices as a stain remover and to make other chemicals. In addition to this, it is used in rosin esters, drying oils, printing inks and flavoring agent.

Solubility
Soluble in water, ethanol and concentrated sulfuric acid. Slightly soluble in ethyl ether and acetone. Insoluble in chloroform, olive oil, carbon tetrachloride, xylene, molten camphor, liquid ammonia and benzene.

Notes
Incompatible with oxidizing agents, amines and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-17-8
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00002700
InChI Key VZCYOOQTPOCHFL-OWOJBTEDSA-N
Synonym fumaric acid, 2-butenedioic acid, trans-butenedioic acid, allomaleic acid, boletic acid, lichenic acid, tumaric acid, 2e-but-2-enedioic acid, butenedioic acid, trans-2-butenedioic acid
PubChem CID 444972
ChEBI CHEBI:18012
IUPAC Name (E)-but-2-enedioic acid
SMILES OC(=O)\C=C\C(O)=O

Specifications

Melting Point ∼299°C (sublimation)
Density 1.625
pH 2.1
Flash Point 273°C (523°F)
Odor Odorless
Quantity 2500 g
Beilstein 605763
Merck Index 14,4287
Solubility Information Soluble in water,ethanol and concentrated sulfuric acid. Slightly soluble in ethyl ether and acetone. Insoluble in chloroform,olive oil,carbon tetrachloride,xylene,molten camphor,liquid ammonia and benzene.
Formula Weight 116.07
Percent Purity 99%
Chemical Name or Material Fumaric acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.