missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fumaric Acid Monomethyl Ester-d3, TRC
Specifications
Specifications
| Chemical Name or Material | Fumaric Acid Monomethyl Ester-d3 |
| CAS | 1616345-41-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C5H3D3O4 |
| Quantity | 100 mg |
| Synonym | (2E)-2-Butenedioic Acid 1-Methyl Ester-d3, (E)-2-Butenedioic Acid Monomethyl Ester-d3, Fumaric Acid Methyl Ester-d3, Methyl Hydrogen Fumarate-d3, Monomethyl Fumarate-d3 |
| InChI Formula | InChI=1S/C5H6O4/c1-9-5(8)3-2-4(6)7/h2-3H,1H3,(H,6,7)/b3-2+/i1D3 |
| SMILES | OC(/C=C/C(OC([2H])([2H])[2H])=O)=O |
| IUPAC Name | (E)-4-(methoxy-d3)-4-oxobut-2-enoic acid |
| Molecular Weight (g/mol) | 133.12 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?