missing translation for 'onlineSavingsMsg'
Learn More

Fumaric Acid, TRC

Product Code. 30003486
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003486 500 mg 500mg
17944935 1 g 1g
17974715 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003486 Supplier Toronto Research Chemicals Supplier No. TRCF500380500MG
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-17-8
Molecular Formula C4 H4 O4
Molecular Weight (g/mol) 116.07
Synonym (E)-Butenedioic Acid, Fumaric Acid, Malic Acid Imp. A (EP), Aspartic Acid Imp. B (EP), Imp. A (EP), 2-Butenedioic acid (E)-, Fumaric acid (8CI), (2E)-But-2-enedioic acid, (E)-2-Butenedioic acid, 2-(E)-Butenedioic acid, 2-Butenedioic acid, (E)-, Allomaleic acid, Bakeshure 451, Bakeshure 470, Boletic acid, FC 33, FC 33 (acid), Lichenic acid, NSC 2752, Petrom, trans-1,2-Ethylenedicarboxylic acid, trans-2-Butenedioic acid, trans-Butenedioic acid
IUPAC Name (E)-but-2-enedioic acid
SMILES OC(=O)\C=C\C(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1+
Formula Weight 116.011
Percent Purity >95%
Chemical Name or Material Fumaric acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.