Learn More
SCAT™ GL45 Adapter for Funnel
Product Code. 17893510 Shop All SCAT Europe Products
Change view
Click to view available options
Closure Size:
S60, GL45
Unit Size:
Each
Product Code. Closure Size Unit Size
17893510 S60, GL45 Each
1 options
Product Code. 17893510 Supplier SCAT™ Supplier No. 107889
  • ONLINE EXCLUSIVE PRICE
    266.00 €
    242.58 € / Pack of 1
    Save 23.42 €
    8% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Robust SCAT™ 'ARNOLD' Safety Funnels with adapters and mechanical level controls for use on various drums with different thread types. Also available in electrically dissipative variants for the safe disposal of explosive liquid waste.
  • Safety funnels based on the SCAT™ 'ARNOLD' funnel series
  • Ready-to-use sets consisting of safety funnels and barrel adapters
  • Made of high-quality plastics for durable and long-lasting use
  • Adapters can be combined with all GL45 safety funnels of the 'ARNOLD' and 'MARCO' series
  • Available for all common thread types and sizes
  • Also available in electrically conductive material for hazardous areas
Specifications
Closure Size S60, GL45
Capacity (Metric) 200 L
Capacity (English) 52.8 gal.
Includes Adapter, Mechanical Level Control
For Use With (Equipment) SCAT ARNOLD Funnel
Material HDPE-EC
Product Type Funnel Adapter
Product Title
Select an issue
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.