missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ Funnel ARNOLD, Ball-Valve

Product Code. 17152245 Shop All SCAT Europe Products
Change view
Click to view available options
Thread Size (Metric):
B53
B83
GL45
S50
S51
S55
S60/61
S65
S70/71
S90
S95
Unit Size:
Each
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) Unit Size
17152245 S55 Each
17182235 B53 Each
17172245 B83 Each
17122245 GL45 Each
17192245 S50 Each
17142245 S51 Each
17112245 S60/61 Each
17162245 S65 Each
17182245 S70/71 Each
17132245 S90 Each
31454223 S95 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 17152245 Supplier SCAT™ Supplier No. 317645
  • ONLINE EXCLUSIVE PRICE
    172.00 €
    156.31 € / Pack of 1
    Save 15.69 €
    9% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The strong safety funnel ARNOLD V2.0 with lid and ball meets the highest requirements for safe disposal of hazardous liquid waste due to its combination of two independent safety mechanisms.

The safety funnel ARNOLD V2.0 with lid and ball valve is available for different container threads. In order to be able to produce in an environmentally friendly and economical way, we supply individual variants with a thread adapter, which has no influence on the functionality or safety of the funnel.

  • Two safety features consist of a ball integrated in the funnel basin and a sealing ring incorporated in the hinged lid of the funnel, which prevent the escape of toxic or explosive gases
  • Made of high quality HDPE material for maximum stability and durability
  • Hinged lid with sealing ring for closure when the funnel is not in use
  • Optimized sieve for catching larger particles and for depositing laboratory bottles
  • Improved splash guard with integrated air channels to prevent bubble formation
  • Ball valve with automatic opening mechanism during filling
  • Individually shortenable safety lance for clean drainage of liquid waste
  • All essential components are removable for easy cleaning

Specifications

Diameter (Metric)Top 200 mm (Funnel)
Capacity (Metric) 1500 mL
Material HDPE
Height (Metric) 210 cm
Autoclavable Not Autoclavable
Color Blue, White
For Use With (Application) Safe Disposal of Liquid and Volatile Wastes
For Use With (Equipment) Disposal Containers
Includes Lance (220 mm), Lid, Ball Valve, Splash Guard, Removable Sieve
Thread Size (Metric) S55
Product Type Safety Funnel
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.