missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ Adapter for Drum Funnel with Mechanical Level Control

Product Code. 17828733 Shop All SCAT Europe Products
Change view
Click to view available options
Material:
Polypropylene
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
17828733 Polypropylene Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17828733 Supplier SCAT™ Supplier No. 107892
  • ONLINE EXCLUSIVE PRICE
    250.00 €
    228.27 € / Pack of 1
    Save 21.73 €
    8% Off
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Level control with screw-in thread, e.g. for drums or canisters. With integrated level indicator and additional threaded connection for mounting a SCAT safety funnel with GL45 thread.

  • Adapter for funnel, mechanical level control, barrel double thread R2" BSP/G2" (m) + BCS 56x4 (m)
  • Thread on top: GL45 (m)
  • Thread on bottom: R2"BSP/G2"(m) + BCS 56x4 (m)
  • Made of high-quality plastics for durable and long-lasting use
  • Adapters can be combined with all GL45 safety funnels of the 'ARNOLD' and 'MARCO' series

Specifications

Capacity (Metric) 200 L
Closure Type R2 BSP/G2 (m) and BCS 56 x 4 (m) to GL45 (m)
Capacity (English) 52.8 gal.
Color Gray
Includes ARNOLD Funnel Adapter, Mechanical Level Control
For Use With (Equipment) SCAT ARNOLD Funnel
Material Polypropylene
Product Type Level Control
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.