missing translation for 'onlineSavingsMsg'
Learn More

Furan-2-boronic acid pinacol ester, 96%

Product Code. 11304985
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11304985 1 g 1g
11314985 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11304985 Supplier Thermo Scientific Alfa Aesar Supplier No. L18366.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 374790-93-9 | C10H15BO3 | 194.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 374790-93-9
Molecular Formula C10H15BO3
Molecular Weight (g/mol) 194.04
MDL Number MFCD02094008
InChI Key GWEWQKZABZXLJH-UHFFFAOYSA-N
Synonym 2-furanboronic acid pinacol ester, furan-2-boronic acid pinacol ester, 2-2-furanyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-furan-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, furan-2-boronic acid pinacol cyclic ester, 2-2-furyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2'-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl furan, 1,3,2-dioxaborolane, 2-2-furanyl-4,4,5,5-tetramethyl, zlchem 693, pubchem18440
PubChem CID 11356013
IUPAC Name 2-(furan-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=CO1

Specifications

Refractive Index 1.472
Quantity 1 g
Beilstein 9045028
Formula Weight 194.04
Percent Purity 98%
Chemical Name or Material Furan-2-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.