missing translation for 'onlineSavingsMsg'
Learn More

Furaneol (technical grade), TRC

Product Code. 30386815
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30386815 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386815 Supplier Toronto Research Chemicals Supplier No. TRCD47102025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,5-Dimethyl-4-hydroxy-3(2H)-furanone (Technical Grade)
CAS 3658-77-3
Molecular Formula C6H8O3
Quantity 25 g
Synonym 2,5-Dimethyl-4-hydroxy-3(2H)-furanone (Technical Grade), 4-Hydroxy-2,5-dimethyl-3(2H)-furanone, 2,5-Dimethyl-3-hydroxy-4-oxo-4,5-dihydrofuran, 2,5-Dimethyl-4,5-dihydrofuran-3-ol-4-one, 2,5-Dimethyl-4-hydroxy-2,3-dihydrofuran-3-one, 4-Hydroxy-2,5-dimethyl-3-oxo-2,3-dihydrofuran, 4-Hydroxy-2,5-dimethyl-2,3-dihydrofuran-3-one, 4-Hydroxy-2,5-dimethyl-2H-furan-3-one, 4-Hydroxy-2,5-dimethyl-3(2H)-furanone, Alletone, Furaneol, Pineapple Ketone
InChI Formula InChI=1S/C6H8O3/c1-3-5(7)6(8)4(2)9-3/h3,8H,1-2H3
SMILES CC1OC(=C(O)C1=O)C
IUPAC Name 4-hydroxy-2,5-dimethylfuran-3-one
Molecular Weight (g/mol) 128.13
Formula Weight 128.0473
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.