missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Furazolidone Discs

Product Code. 11992992
Change view
Click to view available options
Quantity:
50 Discs/Cartridge (250 Discs/Pk.)
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11992992 50 Discs/Cartridge (250 Discs/Pk.) Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11992992 Supplier Thermo Scientific™ Supplier No. DD0028B
  • ONLINE EXCLUSIVE PRICE
    41.55 €
    40.52 € / Pack of 250
    Save 1.03 €
    2% Off
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Differentiate micrococci and staphylococci using these diagnostic discs.

Differentiate micrococci and staphylococci using Thermo Scientific™ Oxoid™ Furazolidone Discs. Furazolidone is a nitrofuran antibacterial agent. Furazilidone susceptibility testing is a reliable method for differentiation of micrococci (resistant) and staphylococci (sensitive).1

Staphylococcus and Micrococcus are Gram-positive, non-motile, and non-sporing and catalyse positive cocci. Both of these microorganisms form the normal flora of skin and mucous membranes of man and animals, the members of the genus Staphylococcus are associated with the clinical infections, the members of Micrococcus are rarely involved in infections.
Use Furazolidone Discs as a reliable and rapid method for differentiation of micrococci and staphylococci.
  • Rapid results
  • Reliable results
  • Simple and ready-to-use cartridge format

Not all products are available for sale in all territories. Please inquire. Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C to +8°C
Description Furazolidone Discs
Detectable Analytes Staphylococcus species
Format Disk, Cartridge
Type Furazolidone Disk
Packaging Type 5 Cart./Pk.
Quantity 50 Discs/Cartridge (250 Discs/Pk.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.