missing translation for 'onlineSavingsMsg'
Learn More

Furazolidone, MP Biomedicals™

Product Code. 11434812 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434812 1 g 1g
11444812 5 g 5g
11454812 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434812 Supplier MP Biomedicals Supplier No. 0219346501
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 67-45-8
Molecular Formula C8H7N3O5
Molecular Weight (g/mol) 225.16
InChI Key PLHJDBGFXBMTGZ-WEVVVXLNSA-N
Synonym furazolidone, furoxone, nitrofurazolidone, furazolidine, nitrofuroxon, furazolidon, diafuron, furaxone, furazol, furazon
PubChem CID 5323714
IUPAC Name 3-[(E)-(5-nitrofuran-2-yl)methylideneamino]-1,3-oxazolidin-2-one
SMILES C1COC(=O)N1N=CC2=CC=C(O2)[N+](=O)[O-]

Specifications

Color Yellow
Quantity 1 g
Formula Weight 225.2
Physical Form Powder
Chemical Name or Material Furazolidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.