missing translation for 'onlineSavingsMsg'
Learn More

Furazolidone, TRC

Product Code. 17962045
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17962045 10 g 10g
17942075 100 mg 100mg
17934685 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17962045 Supplier Toronto Research Chemicals Supplier No. TRCF86410010G
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-45-8
Molecular Formula C8 H7 N3 O5
Molecular Weight (g/mol) 225.16
Synonym Furazolidone, 2-Oxazolidinone, 3-[[(5-nitro-2-furanyl)methylene]amino]-, 2-Oxazolidinone, 3-[(5-nitrofurfurylidene)amino]- (8CI), 3-(5-aci-Nitro-2(5H)-furylidenemethylimino)-2-oxooxazolidinium hydroxide, inner salt (6CI), 3-[[(5-Nitro-2-furanyl)methylene]amino]-2-oxazolidinone, 2-Furanmethanimine, 5-nitro-N-(2-oxo-3-oxazolidinyl)-, 3-[(5-Nitrofurfurylidene)amino]-2-oxazolidinone, Dependal-M, Diafuron, Foroxone, Furaxon, Furaxone, Furazol, Furazolidinone, Furazolidon, Furazolidone, Furazon, Furidon, Furovag, Furox, Furoxal, Furoxane, Furoxon, Furoxone, Furoxone Swine Mix, Furozolidine, Giardil, Giarlam, Medaron, N-(5-Nitro-2-furfurylidene)-3-amino-2-oxazolidone, NF 180, NSC 6469, Neftin, Nicolen, Nifulidone, Nitrofurazolidone, Nitrofuroxon, Optazol, Ortazol, Puradin, Roptazol, Tikofuran, Trifurox
IUPAC Name 3-[(E)-(5-nitrofuran-2-yl)methylideneamino]-1,3-oxazolidin-2-one
SMILES [O-][N+](=O)c1oc(\C=N\N2CCOC2=O)cc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H7N3O5/c12-8-10(3-4-15-8)9-5-6-1-2-7(16-6)11(13)14/h1-2,5H,3-4H2/b9-5+
Formula Weight 225.0386
Chemical Name or Material Furazolidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.