missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl Alcohol, TRC

Product Code. 30133537
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Unit Size:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133537 50 g 50g
30133782 250 g 250g
18053240 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133537 Supplier Toronto Research Chemicals Supplier No. TRCF86422050G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-00-0
Molecular Formula C5 H6 O2
Molecular Weight (g/mol) 98.1
Synonym 2-Hydroxymethylfuran, 2-Furylmethanol, 2-Furfurylalcohol, 2-Furanmethanol, Furfuryl alcohol (8CI), 2-(Hydroxymethyl)furan, 2-Furancarbinol, 2-Furanylmethanol, 2-Furfuryl alcohol, 2-Furylcarbinol, 2-Furylmethyl alcohol, 5-Hydroxymethylfuran, Chem-Rez 200, Chem-Rez Fury 454, Furfuralcohol, Furfuranol, Furylcarbinol, NSC 8843, α-Furfuryl alcohol, α-Furylcarbinol
IUPAC Name furan-2-ylmethanol
SMILES OCc1occc1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C5H6O2/c6-4-5-2-1-3-7-5/h1-3,6H,4H2
Formula Weight 98.0368
Chemical Name or Material Furfuryl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.